Learn More
Biomatik Corporation Human Complexin 2 (CPLX2) ELISA Kit
Supplier:  Biomatik Corporation EKU10747
Encompass_Preferred
Human Complexin 2 (CPLX2) ELISA Kit for detecting CPLX2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6PUV4. Gene ID: 10814. Target Synonyms: 921-L; CPX-2; CPX2; Synaphin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4533