Learn More
Biomatik Corporation Human Conserved Helix Loop Helix Ubiquitous Kinase (CHUK) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10917
Encompass_Preferred
Human Conserved Helix Loop Helix Ubiquitous Kinase (CHUK) High-sensitive ELISA Kit for detecting CHUK in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 1.11pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15111. Gene ID: 1147. Target Synonyms: TCF16; IKK1; IKK-alpha; IkBKA; NFKBIKA; IKKA; Inhibitor of nuclear factor kappa-B kinase subunit alpha; Transcription factor 16; I-kappa-B kinase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8268