Learn More
Biomatik Corporation Human Contactin 6 (CNTN6) ELISA Kit
Supplier:  Biomatik Corporation EKU10749
Encompass_Preferred
Human Contactin 6 (CNTN6) ELISA Kit for detecting CNTN6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UQ52. Gene ID: 27255. Target Synonyms: NB-3; Neural Adhesion Molecule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4528