Learn More
Raybiotech Inc Human/Mouse/Rat Copeptin EIA Kit.
Supplier:  Raybiotech Inc EIACOP1
Encompass_Preferred
Optimized for serum and cell culture supernatants. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8810