Learn More
Sino Biological Anti-COQ7 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12195MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human COQ7 (rh COQ7; Catalog#12195-H08H; Q99807-1; Ser37-Leu217). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2429