Learn More
Sino Biological Anti-COQ7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12195T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human COQ7 (rh COQ7; Catalog#12195-H08H; Q99807-1; Ser37-Leu217). COQ7 specific IgG was purified by Human COQ7 affinity chromatography.
Catalog No. 50-241-2548