Learn More
Sino Biological Anti-ADCK3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102492T4020
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CABC1/ADCK3, and purified by antigen affinity chromatography.
Catalog No. 50-240-8141