Learn More
STEMCELL Technologies Human Cord Blood CD14+ Monocytes, Frozen, Size: 5 x 10^6 cells
SDP
Catalog No. 501975787
Encompass
Primary human CD14+ monocytes were isolated from umbilical cord blood (CB) mononuclear cells (MNCs) using positive immunomagnetic separation techniques. CB was collected using citrate-phosphate-dextrose (CPD) as an anticoagulant. CD14 is primarily expressed on monocytes.Cells were collected using consent forms and protocols approved by either an Institutional Review Board, the Food and Drug Administration, the U.S. Department of Health and Human Services, and/or an equivalent regulatory authority.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.
Catalog No. 50-197-5787 Supplier STEMCELL Technologies Supplier No. 70018
Add to Cart
Add to Cart
Primary human CD14+ monocytes were isolated from umbilical cord blood (CB) mononuclear cells (MNCs) using positive immunomagnetic separation techniques. CB was collected using citrate-phosphate-dextrose (CPD) as an anticoagulant. CD14 is primarily expressed on monocytes.Cells were collected using consent forms and protocols approved by either an Institutional Review Board, the Food and Drug Administration, the U.S. Department of Health and Human Services, and/or an equivalent regulatory authority.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.