Learn More
STEMCELL Technologies Human Cord Blood Mononuclear Cells, Frozen, Size: 1.5 x 10^8 cells
SDP
Supplier:  STEMCELL Technologies 70007
Encompass
Streamline your assays with ready-to-use, ethically sourced, primary mononuclear cells, including lymphocytes, monocytes, hematopoietic stem and progenitor cells, and mesenchymal stromal cells. With personalized service, custom products, flexible delivery times, and the option to reserve entire lots to prescreen cells for applications, we help you get the cells you need. Isolated from umbilical cord blood and cryopreserved in animal component-free CryoStorCS10 medium, cells are collected using Institutional Review Board (IRB) or other regulatory authority-approved consent forms and protocols. Additional documentation and high-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. CPD is added as an anticoagulant. Contact techsupport@stemcell.com for donor specifications (BMI category, smoking status, ethnicity, etc.) Donors are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C. Certain products are only available in select territories.
Catalog No. NC1569791