Learn More
STEMCELL Technologies Human Cord Blood Pan-T Cells, Frozen, Size: 1.5 x 10^7 cells
SDP
Catalog No. 501975784
Encompass
Primary human pan-T cells were isolated from umbilical cord blood (CB) mononuclear cells (MNCs) using negative immunomagnetic separation techniques. CB was collected using citrate-phosphate-dextrose (CPD) as an anticoagulant.Cells were collected using consent forms and protocols approved by either an Institutional Review Board, the Food and Drug Administration, the U.S. Department of Health and Human Services, and/or an equivalent regulatory authority.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.
Catalog No. 50-197-5784 Supplier STEMCELL Technologies Supplier No. 70014
Primary human pan-T cells were isolated from umbilical cord blood (CB) mononuclear cells (MNCs) using negative immunomagnetic separation techniques. CB was collected using citrate-phosphate-dextrose (CPD) as an anticoagulant.Cells were collected using consent forms and protocols approved by either an Institutional Review Board, the Food and Drug Administration, the U.S. Department of Health and Human Services, and/or an equivalent regulatory authority.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/primary-cells-faq) on Primary Cells.