Learn More
STEMCELL Technologies Human Cord Blood Plasma, Frozen, Size: 20 mL
SDP
Catalog No. 501975791
Encompass
Human plasma has been isolated from umbilical cord blood (CB) using centrifugation. CB was collected using citrate-phosphate-dextrose (CPD) as an anticoagulant.Plasma was collected using consent forms and protocols approved by either an Institutional Review Board, the Food and Drug Administration, the U.S. Department of Health and Human Services, and/or an equivalent regulatory authority.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
Catalog No. 50-197-5791 Supplier STEMCELL Technologies Supplier No. 70020.2
Human plasma has been isolated from umbilical cord blood (CB) using centrifugation. CB was collected using citrate-phosphate-dextrose (CPD) as an anticoagulant.Plasma was collected using consent forms and protocols approved by either an Institutional Review Board, the Food and Drug Administration, the U.S. Department of Health and Human Services, and/or an equivalent regulatory authority.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.