Learn More
Biomatik Corporation Human Core Binding Factor Beta Subunit (CBFb) ELISA Kit
Supplier:  Biomatik Corporation EKU12331
Encompass_Preferred
Human Core Binding Factor Beta Subunit (CBFb) ELISA Kit for detecting CBFb in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13951. Gene ID: 865. Target Synonyms: CBF-B; PEBP2B; Polyomavirus enhancer-binding protein 2 beta subunit; SL3-3 enhancer factor 1 subunit beta; SL3/AKV core-binding factor beta subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7982