Learn More
ATCC Human coronavirus 229E; RNA; Strain: 229E
Catalog No. 502382053
Encompass
Genomic RNA isolated from a preparation of cell lysate and supernatant from MRC-5 cells (ATCC CCL-171) infected with Human coronavirus 229E strain 229E (ATCC VR-740). This product was prepared using methods known to inactivate viruses. This product can be used in PCR and other molecular biology applications. Source organism and host cells are also available through the ATCC Catalog.
Catalog No. 50-238-2053 Supplier ATCC Supplier No. VR740D
Genomic RNA isolated from a preparation of cell lysate and supernatant from MRC-5 cells (ATCC CCL-171) infected with Human coronavirus 229E strain 229E (ATCC VR-740). This product was prepared using methods known to inactivate viruses. This product can be used in PCR and other molecular biology applications. Source organism and host cells are also available through the ATCC Catalog.