Learn More
ATCC Human coronavirus 229E; Strain: 229E
Catalog No. 502382706
Encompass
Human coronavirus 229E is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from the nasal and throat swabs of a man with a mild upper respiratory illness. The strain has applications in respiratory disease research and does not result in hemolysis.
Catalog No. 50-238-2706 Supplier ATCC Supplier No. VR740
Human coronavirus 229E is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from the nasal and throat swabs of a man with a mild upper respiratory illness. The strain has applications in respiratory disease research and does not result in hemolysis.