Learn More
Sino Biological Human coronavirus (HCoV-229E) Nucleocapsid Antibody, Rabbit MAb 100 µL
Catalog No. 5000749816
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human coronavirus (HCoV-229E) Nucleocapsid ( Catalog#40640-V07E; NP_073556.1; Met1-Asn389).
Catalog No. 50-007-49816 Supplier Sino Biological Supplier No. 40640R060100
This antibody was obtained from a rabbit immunized with purified, recombinant Human coronavirus (HCoV-229E) Nucleocapsid ( Catalog#40640-V07E; NP_073556.1; Met1-Asn389).