Learn More
Sino Biological Human coronavirus (HCoV-NL63) Nucleocapsid Antibody, Mouse MAb 20 µL
Catalog No. 5000748810
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human coronavirus (HCoV-NL63) Nucleocapsid ( Catalog#40641-V07E; YP_003771.1; Met1-His377). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-48810 Supplier Sino Biological Supplier No. 40641MM1420
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human coronavirus (HCoV-NL63) Nucleocapsid ( Catalog#40641-V07E; YP_003771.1; Met1-His377). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.