Learn More
Sino Biological Human coronavirus (HCoV-NL63) Nucleocapsid Antibody, Rabbit MAb 20 µL
Catalog No. 5000748942
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human coronavirus (HCoV-NL63) Nucleocapsid ( Catalog#40641-V07E; YP_003771.1; Met1-His377).
Catalog No. 50-007-48942 Supplier Sino Biological Supplier No. 40641R00520
This antibody was obtained from a rabbit immunized with purified, recombinant Human coronavirus (HCoV-NL63) Nucleocapsid ( Catalog#40641-V07E; YP_003771.1; Met1-His377).