Learn More
Sino Biological Human coronavirus (HCoV-OC43) Nucleocapsid Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40643T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human coronavirus (HCoV-OC43) Nucleoprotein / NP ( Catalog#40643-V07E; YP_009555245.1; Met1-Ile448). Coronavirus Nucleocapsid specific IgG was purified by Human coronavirus (HCoV-OC43) Nucleoprotein / NP affinity chromatography.
Catalog No. 50-224-7373