Learn More
Sino Biological Human coronavirus HKU1 (isolate N5) (HCoV-HKU1) Spike S1+S2 Protein (ECD, His Tag), Biotinylated
Supplier:  Sino Biological 40606V08BB20
Encompass_Preferred
A DNA sequence encoding the human coronavirus HKU1 (isolate N5) (HCoV-HKU1) spike protein (S1+S2 ECD) (Q0ZME7.1) (Met1-Pro1295) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-224-7130