Learn More
Sino Biological Human coronavirus HKU1 (isolate N1) (HCoV-HKU1) Spike S2 Protein (His Tag) 100ug
Supplier:  Sino Biological 40021V08B100
Encompass_Preferred
A DNA sequence encoding the Human coronavirus HKU1 (isolate N1) (HCoV-HKU1) Spike S2 (YP_173238.1)(Ser761-Pro1300) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6651