Learn More
Brainxell Inc Cortical GABAergic Neurons (Human iPSC-Derived, Male Line, 1M)
SDP
Catalog No. NC2259386
Encompass
Cortical GABAergic neurons derived from iPSC line WC-30, male. Contents: 1 vial of ≥1x10^6 viable cryopreserved cells. Marker Expression: Cortical GABAergic Neurons (BX-0400) have high neuronal purity (>80%) and comprise predominantly inhibitory neurons. Labelling with the pan-neuronal marker MAP2(green) and for GABA (red) highlight the purity of this neuronal product. Function: Cortical GABAergic Neurons exhibit pronounced electrophysiological activity after two weeks in culture, as demonstrated on multi-electrode array (MEA) recording systems. Related diseases: Epilepsy, Alzheimer's Disease, & Schizophrenia. Primary neurotransmitter released: GABA
Catalog No. NC2259386 Supplier Brainxell Inc Supplier No. BX0400301M
Cortical GABAergic neurons derived from iPSC line WC-30, male. Contents: 1 vial of ≥1x10^6 viable cryopreserved cells. Marker Expression: Cortical GABAergic Neurons (BX-0400) have high neuronal purity (>80%) and comprise predominantly inhibitory neurons. Labelling with the pan-neuronal marker MAP2(green) and for GABA (red) highlight the purity of this neuronal product. Function: Cortical GABAergic Neurons exhibit pronounced electrophysiological activity after two weeks in culture, as demonstrated on multi-electrode array (MEA) recording systems. Related diseases: Epilepsy, Alzheimer's Disease, & Schizophrenia. Primary neurotransmitter released: GABA