Learn More
Biomatik Corporation Human Corticotropin Releasing Hormone Binding Protein (CRHBP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11995
Encompass_Preferred
Human Corticotropin Releasing Hormone Binding Protein (CRHBP) Mini sample ELISA Kit for detecting CRHBP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4,000pg/ml. Sensitivity: 22.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24387. Gene ID: 1393. Target Synonyms: CRF-BP; CRFBP; Corticotropin-Releasing Factor-Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5063