Learn More
Crystal Chem Inc Human Cortisol (Saliva) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80957
Encompass
The Cortisol ELISA (Saliva) is a highly sensitive assay that is used to quantify levels of human cortisol in saliva. Highlights: Kit uses sample size of only 50 µL. High sensitivity of 1 ng/mL. Dynamic range of 1 - 100 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7863