Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cot-1 DNA™-Fluorometric QC

Catalog No. 15279101
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-279-101 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-279-101 Supplier Invitrogen™ Supplier No. 15279101
Only null left
Add to Cart
Edge
Add to Cart

Includes

Human Cot-1 DNA™-Fluorometric QC is supplied at 1mg/mL in 10mM Tris-HCl (pH 7.4), 1mM EDTA.

Quantifying by fluorometry provides greater data accuracy

Human Cot-1 DNA-Fluorometric QC is placental DNA predominantly 50 to 300 bp in size and enriched for repetitive DNA sequences, such as the Alu and Kpn family members (1,2). Quantifying Cot-1 DNA™ by fluorometry provides more accurate concentration data of higher molecular weight products, making it beneficial to certain genomic-based array experiments, such as array CGH using BAC arrays. Human Cot-1 DNA™-Fluorometric QC suppresses prevalent repetitive elements during hybridization, improving results.

Array CGH, Cell Analysis, Cellular Imaging, ChIP-on-Chip, Chromatin Biology, Fluorescence In Situ Hybridization, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, In Situ Hybridization (ISH), Microarray Analysis, RNAi, Epigenetics and Non-Coding RNA Research

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type DNA QC
Content And Storage Human Cot-1 DNA™-Fluorometric QC is supplied at 1 mg/ml in 10 mM Tris-HCl (pH 7.4), 1 mM EDTA. Store at -20°C. Guaranteed stable for 12 months when properly stored.
Reagent Type Cot-1 DNA
Shipping Condition Dry Ice
Species Human
Product Line Cot-1 DNA
For Use With (Application) Block Non-Specific Hybridization
Quantity 1 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.