Learn More
Raybiotech Inc Human COX-2 (PTGS2) ELISA Kit
Supplier:  Raybiotech Inc ELHCOX21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 1.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human COX-2. Other species not determined.
Catalog No. 50-194-9145