Learn More
Sino Biological Human COX4I1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 106894T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human COX IV / COX4I1, and purified by antigen affinity chromatography.
Catalog No. 50-237-2882