Learn More
Biomatik Corporation Human Coxsackie And Adenovirus Receptor Like Membrane Protein (CLMP) ELISA Kit
Supplier:  Biomatik Corporation EKU03481
Encompass_Preferred
Human Coxsackie And Adenovirus Receptor Like Membrane Protein (CLMP) ELISA Kit for detecting CLMP in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H6B4. Gene ID: 79827. Target Synonyms: ACAM; ASAM; Adipocyte Adhesion Molecule; Adipocyte-Specific Adhesion Molecule; CXADR Like Membrane Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4969