Learn More
ATCC Human Coxsackievirus A10; Strain: M.K.(Kowalik)
Catalog No. 502383880
Encompass
Human Coxsackievirus A 10 strain M.K. (Kowalik) is propagated in RD cells (ATCC CCL-136). This strain was isolated from the stool of a 7-year-old child in New York in 1950.
Catalog No. 50-238-3880 Supplier ATCC Supplier No. VR168
Human Coxsackievirus A 10 strain M.K. (Kowalik) is propagated in RD cells (ATCC CCL-136). This strain was isolated from the stool of a 7-year-old child in New York in 1950.