Learn More
ATCC Human Coxsackievirus A21; Strain: Kuykendall
Catalog No. 502382399
Encompass
Human Coxsackievirus A 21 strain Kuykendall [V-024-001-012] is propagated in WISH cells (ATCC CCL-25). This strain was isolated in 1952 from the stool of a 17-year-old human male with paralytic illness and was deposited by NIH/NIAID. This product has applications in respiratory disease research.
Catalog No. 50-238-2399 Supplier ATCC Supplier No. VR850
Human Coxsackievirus A 21 strain Kuykendall [V-024-001-012] is propagated in WISH cells (ATCC CCL-25). This strain was isolated in 1952 from the stool of a 17-year-old human male with paralytic illness and was deposited by NIH/NIAID. This product has applications in respiratory disease research.