Learn More
ATCC Human coxsackievirus A6; Strain: Gdula
Catalog No. 502383357
Encompass
Human Coxsackievirus A6 strain Gdula is propagated in RD cells (ATCC CCL-136). This strain was isolated in 1949 in New York from the stool of a 5-year-old child with a sore throat, fever, and leg muscle spasms. It can be used in enteric disease research.
Catalog No. 50-238-3357 Supplier ATCC Supplier No. VR1801
Human Coxsackievirus A6 strain Gdula is propagated in RD cells (ATCC CCL-136). This strain was isolated in 1949 in New York from the stool of a 5-year-old child with a sore throat, fever, and leg muscle spasms. It can be used in enteric disease research.