Learn More
ATCC Human Coxsackievirus B1; Strain: Conn-5
Catalog No. 502382236
Encompass
Human Coxsackievirus B1 strain Conn-5 is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1). This strain was isolated in 1948 from the stool of a human patient with aseptic meningitis in Connecticut, US, and was deposited by the Yale University School of Medicine.
Catalog No. 50-238-2236 Supplier ATCC Supplier No. VR28
Add to Cart
Add to Cart
Human Coxsackievirus B1 strain Conn-5 is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1). This strain was isolated in 1948 from the stool of a human patient with aseptic meningitis in Connecticut, US, and was deposited by the Yale University School of Medicine.