Learn More
ATCC Human Coxsackievirus B3; Strain: Nancy
Catalog No. 502383761
Encompass
Human Coxsackievirus B3 strain Nancy is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1) This strain was isolated in 1949 from the stool of a febrile patient with minor illness in Connecticut.
Catalog No. 50-238-3761 Supplier ATCC Supplier No. VR30
Human Coxsackievirus B3 strain Nancy is propagated in LLC-MK2 Derivative cells (ATCC CCL-7.1) This strain was isolated in 1949 from the stool of a febrile patient with minor illness in Connecticut.