Learn More
ATCC Human Coxsackievirus B5; Strain: Faulkner
Catalog No. 502382350
Encompass
Human Coxsackievirus B 5 strain Faulkner is propagated in LLC-MK2 Original cells (ATCC CCL-7). This strain was isolated in 1952 from a child with suspected poliomyelitis in Kentucky. It has applications in infectious disease research.
Catalog No. 50-238-2350 Supplier ATCC Supplier No. VR185
Human Coxsackievirus B 5 strain Faulkner is propagated in LLC-MK2 Original cells (ATCC CCL-7). This strain was isolated in 1952 from a child with suspected poliomyelitis in Kentucky. It has applications in infectious disease research.