Learn More
Sino Biological Anti-Carboxypeptidase A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10504RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Carboxypeptidase A1 / CPA1 (rh Carboxypeptidase A1 / CPA1; Catalog#10504-H08H; NP_001859.1; Met 1-Tyr 419). Carboxypeptidase A1 / CPA1 specific IgG was purified by Human Carboxypeptidase A1 / CPA1 affinity chromatography.
Catalog No. 50-241-0869