Learn More
Sino Biological Recombinant Anti-Carboxypeptidase A2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10499R00920
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase A2 / CPA2 (rh Carboxypeptidase A2 / CPA2; Catalog#10499-H08H; NP_001859.1; Met1-Tyr417).
Catalog No. 50-240-8834