Learn More
Sino Biological Anti-Carboxypeptidase B1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10501MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Carboxypeptidase B1 / CPB1 (rh Carboxypeptidase B1 / CPB1; Catalog#10501-H08H; NP_001862.2; Met1-Tyr417). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9264