Learn More
Sino Biological Anti-Carboxypeptidase E/CPE Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10069RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CPE (rhCPE; Catalog#10069-H08H; NP_001864.1; Met 1-Ser 453). CPE specific IgG was purified by human CPE affinity chromatography.
Catalog No. 50-241-0069