Learn More
Sino Biological Anti-Carboxypeptidase E/CPE Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10069T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Carboxypeptidase E / CPE (rh Carboxypeptidase E / CPE; Catalog#10069-H08H; NP_001864.1; Met1-Ser453). Carboxypeptidase E / CPE specific IgG was purified by Human Carboxypeptidase E / CPE affinity chromatography.
Catalog No. 50-240-8632