Learn More
Sino Biological Human CPE Protein 50ug (Sino Biological)
Supplier:  Sino Biological 10069H02H50
Encompass_Preferred
A DNA sequence encoding the human carboxypeptidase E (CPE) precursor (NP_001864.1) (Met 1-Ser 453) was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-237-2375