Learn More
Biomatik Corporation Human C-Peptide (CP) CLIA Kit
Supplier:  Biomatik Corporation EKU08617
Encompass_Preferred
Human C-Peptide (CP) CLIA Kit for detecting CP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.1-20,000pg/ml. Sensitivity: 31.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P01308. Gene ID: 3630. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8652