Learn More
Sino Biological Recombinant Anti-CD21 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10811R257A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD21 (rh CD21; Catalog#10811-H08H; P20023-1; Met1-Arg971) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-7746