Learn More
Sino Biological Anti-CD21 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10811RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD21 (rh CD21; Catalog#10811-H08H; P20023-1; Met 1-Arg 971). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1833