Learn More
Sino Biological Anti-CRABP2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11203MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CRABP2 (rh CRABP2; Catalog#11203-H07E; NP_001869.1; Pro2-Glu 138). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2449