Learn More
Sino Biological Recombinant Anti-CRABP2 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 11203R004P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CRABP2 (rh CRABP2; Catalog#11203-H07E; NP_001869.1; Pro 2-Glu 138) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1802