Learn More
Sino Biological Anti-CRABP2 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11203MM05F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CRABP2 (rh CRABP2; Catalog#11203-H07E; NP_001869.1; Pro2-Glu138) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1390