Learn More
Sino Biological Anti-CRABP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11203RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CRABP2 (rh CRABP2; Catalog#11203-H07E; NP_001869.1; Pro2-Glu 138). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2108