Learn More
Biomatik Corporation Human C-Raf Proto Oncogene Serine/Threonine Protein Kinase (Raf-1) ELISA Kit
Supplier:  Biomatik Corporation EKU03485
Encompass_Preferred
Human C-Raf Proto Oncogene Serine/Threonine Protein Kinase (Raf-1) ELISA Kit for detecting Raf-1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04049. Gene ID: 5894. Target Synonyms: RAF1; CRAF; V-raf-1 Murine Leukemia Viral Oncogene Homolog 1; Proto-oncogene c-RAF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9031