Learn More
Sino Biological Recombinant Human CRBN & DDB1 Heterodimer Protein (His & FLAG Tag)
Catalog No. 502993084
Encompass_Preferred
A DNA sequence encoding the Human CRBN (NP_057386.2)(Met1-Leu442) was expressed, with a polyhistidine tag at the N-terminus, constructed the plasmid 1; A DNA sequence encoding the Human DDB1 (NP_001914.3) (Ser2-His1140) was expressed with a flag tag at the N-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the CRBN & DDB1 Heterodimer was purified.
Catalog No. 50-299-3084 Supplier Sino Biological Supplier No. CT141H0722B100
A DNA sequence encoding the Human CRBN (NP_057386.2)(Met1-Leu442) was expressed, with a polyhistidine tag at the N-terminus, constructed the plasmid 1; A DNA sequence encoding the Human DDB1 (NP_001914.3) (Ser2-His1140) was expressed with a flag tag at the N-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the CRBN & DDB1 Heterodimer was purified.