Learn More
Biomatik Corporation Human Creatine Kinase B (CK-BB) CLIA Kit
Supplier:  Biomatik Corporation EKU09897
Encompass_Preferred
Human Creatine Kinase B (CK-BB) CLIA Kit for detecting CK-BB in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.27-200ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P12277. Gene ID: 1152. Target Synonyms: CKB; CKBB; CK-B; CK-1; Creatine Kinase, Brain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6008