Learn More
Biomatik Corporation Human Creatine Transporter (CRT) ELISA Kit
Supplier:  Biomatik Corporation EKU03496
Encompass_Preferred
Human Creatine Transporter (CRT) ELISA Kit for detecting CRT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48029. Gene ID: 6535. Target Synonyms: SLC6A8; CT1; CRTR; Sodium And Chloride Dependent Creatine Transporter 1; Solute Carrier Family 6 Member 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4737